Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il31ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139299.2 | NP_647460.2 | 9 | 1102 | ||
GenBank mRNA | AB083111.1 | - | 9 | 1101 | ||
AF486621.1 | - | 6 | 681 | |||
AY509150.1 | - | 998 | ||||
AY509151.1 | - | 998 | ||||
BC066164.1 | - | 7 | 823 | 30475340 | ||
BC145083.1 | - | 8 | 1005 | |||
BC145084.1 | - | 8 | 1005 | |||
BC145085.1 | - | 9 | 1128 |