Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290543.1 | NP_001277472.1 | 4, 5 | 578 | ||
NM_139306.3 | NP_647467.1 | 4, 5 | 578 | |||
GenBank mRNA | AF282864.1 | - | 4, 5 | 537 | ||
AK085306.1 | - | 4, 5 | 574 | |||
AY312515.1 | - | 4, 5 | 503 |