Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159415.1 | NP_001152887.1 | 8 | 955 | ||
NM_144511.2 | NP_653094.2 | 8 | 955 | |||
GenBank mRNA | BC010812.1 | - | 8 | 945 | ||
BC019147.1 | - | 8 | 951 | 5052937 | ||
BC019926.1 | - | 8 | 946 | |||
CB946634.1 | - | 2 | 131 | |||
CB947075.1 | - | 2 | 131 |