Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tifa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145133.3 | NP_660115.1 | 3 | 618 | ||
GenBank mRNA | AB062111.1 | - | 3 | 415 | ||
AK041891.1 | - | 2 | 452 | |||
AK044221.1 | - | 2 | 562 | |||
AK149408.1 | - | 3 | 454 | |||
AK149864.1 | - | 3 | 586 | |||
AK151651.1 | - | 3 | 569 | |||
AK166239.1 | - | 3 | 660 | |||
BC065775.1 | - | 3 | 613 | 1528121 |