Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005744.3 | NP_005735.2 | 2, 3 | 765 | ||
XM_011521439.2 | XP_011519741.1 | 3, 4 | 279 | |||
GenBank mRNA | AB209432.1 | - | 3, 4 | 394 | ||
AF072832.1 | - | 2, 3 | 451 | |||
AJ130976.1 | - | 2, 3 | 169 | |||
AJ243190.1 | - | 2, 3 | 765 | |||
AK312715.1 | - | 2, 3 | 764 | |||
BC051877.1 | - | 2, 3 | 700 | 5742498 |