Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146238.3 | NP_666350.2 | 4 | 1148 | ||
GenBank mRNA | AK047511.1 | - | 5 | 1243 | ||
AK053774.1 | - | 4 | 1112 |