Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cchcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146248.2 | NP_666360.2 | 18 | 2362 | ||
GenBank mRNA | AK159780.1 | - | 18 | 2289 | ||
BC031416.1 | - | 19 | 2451 | 4012768 | ||
CJ238541.1 | - | 5 | 410 |