Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277229.1 | NP_001264158.1 | 2 | 728 | ||
NM_001277230.1 | NP_001264159.1 | 2 | 728 | |||
NM_153398.3 | NP_700447.2 | 2 | 728 | |||
GenBank mRNA | AF263010.2 | - | 2 | 651 | ||
AK054097.1 | - | 2 | 684 | |||
AK146986.1 | - | 2 | 704 | |||
AK154467.1 | - | 2 | 704 | |||
AK169260.1 | - | 2 | 675 | |||
AY210402.1 | - | 2 | 702 | |||
BC050933.1 | - | 2 | 690 | 6811382 | ||
BC055367.1 | - | 2 | 671 | 30056072 |