Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153572.2 | NP_705800.1 | 8 | 1059 | ||
GenBank mRNA | AK049620.1 | - | 8 | 1413 | ||
BC030434.1 | - | 8 | 1046 | 5369313 |