Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172113.1 | NP_001165584.1 | 10, 11 | 1218 | ||
NM_001172114.1 | NP_001165585.1 | 9, 10 | 1098 | |||
NM_153792.2 | NP_722487.2 | 10, 11 | 1238 | |||
GenBank mRNA | AK088431.1 | - | 9, 10 | 1007 | ||
AK131617.1 | - | 10, 11 | 1179 | |||
BC008598.1 | - | 10, 11 | 1141 | 3499933 |