Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172113.1 | NP_001165584.1 | 16 | 1620 | ||
NM_001172114.1 | NP_001165585.1 | 15 | 1500 | |||
NM_153792.2 | NP_722487.2 | 16 | 1640 | |||
GenBank mRNA | AK088431.1 | - | 15 | 1409 | ||
AK131617.1 | - | 16 | 1581 | |||
AK178583.1 | - | 1 | 51 | |||
BC005649.1 | - | 4 | 252 | |||
BC008598.1 | - | 16 | 1543 | 3499933 | ||
BC031414.1 | - | 2 | 1445 |