Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153799.3 | NP_722494.1 | 7 | 2901 | ||
GenBank mRNA | AK090005.1 | - | 2594 | |||
AK146804.1 | - | 7 | 2931 | |||
AK166286.1 | - | 7 | 2885 | |||
AK207836.1 | - | 1 | 210 | |||
BC031725.1 | - | 7 | 2888 | 4017604 | ||
BC033484.1 | - | 7 | 2902 | 4954674 |