Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crocc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145958.1 | NP_001139430.1 | 31 | 4921 | ||
NM_172122.2 | NP_742120.2 | 32 | 5174 | |||
GenBank mRNA | AF527975.1 | - | 32 | 5174 | ||
AK032335.1 | - | 3 | 839 | |||
AK122285.1 | - | 23 | 4417 | |||
BC027090.1 | - | 5 | 715 | |||
BC034536.1 | - | 9 | 2079 | |||
BC036729.1 | - | 17 | 2816 | |||
BC038074.1 | - | 1 | 56 | |||
BC054054.1 | - | 30 | 4902 | 6394697 |