Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172535.3 | NP_766123.2 | 9 | 1587 | ||
GenBank mRNA | AK029931.1 | - | 9 | 1577 | ||
AK133019.1 | - | 9 | 1569 | |||
AK161501.1 | - | 3 | 412 | |||
BC137937.1 | - | 9 | 1554 | 8860956 | ||
BC137957.1 | - | 9 | 1554 | 8860976 |