Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h12d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172785.3 | NP_766373.2 | 6 | 2049 | ||
GenBank mRNA | AK052806.1 | - | 6 | 2051 | ||
BC112398.1 | - | 5 | 1831 | 40062049 |