Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172840.2 | NP_766428.2 | 10 | 1118 | ||
GenBank mRNA | AJ536329.1 | - | 10 | 1118 | ||
AK029297.1 | - | 10 | 1065 | |||
AK133486.1 | - | 10 | 1025 | |||
AK160971.1 | - | 10 | 1124 | |||
BC115868.1 | - | 9 | 1020 | 40081354 | ||
BC116636.1 | - | 10 | 1068 | 40081359 |