Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172840.2 | NP_766428.2 | 11 | 1188 | ||
GenBank mRNA | AJ536329.1 | - | 11 | 1188 | ||
AK029297.1 | - | 11 | 1135 | |||
AK133486.1 | - | 11 | 1095 | |||
AK145058.1 | - | 11 | 1358 | |||
AK160971.1 | - | 11 | 1194 | |||
AK166339.1 | - | 11 | 1266 | |||
BC115868.1 | - | 10 | 1090 | 40081354 | ||
BC116636.1 | - | 11 | 1138 | 40081359 |