Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172840.2 | NP_766428.2 | 14 | 3319 | ||
GenBank mRNA | AK029297.1 | - | 14 | 3265 | ||
AK160971.1 | - | 14 | 3327 |