Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094608.1 | - | 2301 |
Gene Symbol: C230030N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082264.1 | - | 2600 |