Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankmy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172850.3 | NP_766438.2 | 5, 6 | 1046 | ||
GenBank mRNA | AK029700.1 | - | 5, 6 | 1048 | ||
BC120887.1 | - | 4, 5 | 882 | 40060916 | ||
BC120888.1 | - | 4, 5 | 882 | 40060917 |