Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntnap5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081653.1 | NP_001075122.1 | 13 | 2003 |
Gene Symbol: Cntnap5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172851.2 | NP_766439.2 | 13 | 2003 | ||
GenBank mRNA | AK082656.1 | - | 7 | 1061 | ||
AK133394.1 | - | 3 | 333 |