Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172861.3 | NP_766449.1 | 8 | 3345 | ||
GenBank mRNA | AK173217.1 | - | 8 | 3321 | ||
BC061928.1 | - | 8 | 3321 | 5359765 |