Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201359.1 | NP_001188288.1 | 6 | 944 | ||
NM_005624.3 | NP_005615.2 | 6 | 947 | |||
XM_011528177.2 | XP_011526479.1 | 7 | 1148 | |||
XM_017027119.1 | XP_016882608.1 | 7 | 1114 | |||
XM_017027120.1 | XP_016882609.1 | 7 | 1043 | |||
XM_017027121.1 | XP_016882610.1 | 7 | 989 | |||
XM_017027122.1 | XP_016882611.1 | 7 | 1123 |