Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp8nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173031.3 | NP_766619.2 | 5, 6 | 499 | ||
GenBank mRNA | AK028610.1 | - | 5, 6 | 501 | ||
BC066135.1 | - | 5, 6 | 455 | |||
BC150940.1 | - | 5, 6 | 454 | |||
BC172093.1 | - | 5, 6 | 454 |