Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp8nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173031.3 | NP_766619.2 | 7, 8 | 701 | ||
GenBank mRNA | AK028610.1 | - | 7, 8 | 703 | ||
BC150940.1 | - | 7, 8 | 656 | |||
BC172093.1 | - | 7, 8 | 656 |