Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leprot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175036.4 | NP_778201.1 | 4 | 1520 | ||
GenBank mRNA | AK009569.1 | - | 4 | 1521 | ||
AK088848.1 | - | 2 | 1337 | |||
AK152008.1 | - | 4 | 1586 | |||
AK159489.1 | - | 4 | 1591 | |||
AK168597.1 | - | 4 | 1557 | |||
BC004744.1 | - | 4 | 1531 | 3583760 | ||
BC010289.1 | - | 4 | 1449 | 2651399 |