Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289654.1 | NP_001276583.1 | 3 | 1284 | ||
NM_001289655.1 | NP_001276584.1 | 3 | 1284 | |||
NM_175276.4 | NP_780485.2 | 3 | 1284 | |||
GenBank mRNA | AB078608.1 | - | 3 | 462 | ||
AB078609.1 | - | 3 | 462 | |||
AB100088.1 | - | 3 | 462 | |||
AK046161.1 | - | 3 | 1286 | |||
AK148245.1 | - | 3 | 426 | |||
BC138046.1 | - | 3 | 384 | 8861065 |