Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175380.5 | NP_780589.3 | 3, 4 | 634 | ||
GenBank mRNA | AK050572.1 | - | 3, 4 | 527 | ||
AK075845.1 | - | 3, 4 | 476 | |||
AK145475.1 | - | 3, 4 | 489 | |||
AK166662.1 | - | 3, 4 | 487 | |||
AK172886.1 | - | 3, 4 | 579 | |||
BC037729.1 | - | 3, 4 | 497 | |||
BC138976.1 | - | 3, 4 | 462 | 8861998 | ||
BC138980.1 | - | 3, 4 | 462 | 8862002 |
Gene Symbol: 1700022A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006230.1 | - | 2 | 634 |