Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175649.5 | NP_783580.1 | 8 | 628 | ||
GenBank mRNA | AK142511.1 | - | 689 | |||
AK154666.1 | - | 8 | 739 | |||
AY165628.1 | - | 8 | 628 | |||
BC111108.1 | - | 8 | 616 | |||
BC138819.1 | - | 8 | 677 | 8861841 | ||
BC138820.1 | - | 8 | 678 | 8861842 | ||
BC152357.1 | - | 8 | 624 | |||
BC152358.1 | - | 8 | 628 | 40055748 |