Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btn2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289614.1 | NP_001276543.1 | 7 | 2195 | ||
NM_001289615.1 | NP_001276544.1 | 8 | 2163 | |||
NM_175938.3 | NP_787952.2 | 8 | 2222 | |||
GenBank mRNA | AK051061.1 | - | 8 | 2211 | ||
AY177686.1 | - | 10 | 2079 |