Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ush1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176847.3 | NP_789817.1 | 2 | 979 | ||
GenBank mRNA | AB087502.1 | - | 2 | 979 | ||
BC120509.1 | - | 2 | 962 | 40129432 | ||
BC137807.1 | - | 2 | 962 | 8860826 |