Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177128.4 | NP_796102.2 | 10 | 1073 | ||
GenBank mRNA | AK078554.1 | - | 10 | 1066 | ||
AK087791.1 | - | 2 | 2255 | |||
AK165220.1 | - | 10 | 1057 | |||
BC029084.1 | - | 10 | 1013 | 3157011 |