Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177150.2 | NP_796124.1 | 2 | 219 | ||
GenBank mRNA | AK089172.1 | - | 2 | 266 | ||
AK090340.1 | - | 2 | 220 | |||
AK143613.1 | - | 2 | 182 | |||
AK167376.1 | - | 2 | 220 | |||
AK180349.1 | - | 2 | 98 | |||
AK186671.1 | - | 2 | 98 | |||
AK187972.1 | - | 2 | 98 | |||
AK189344.1 | - | 2 | 98 | |||
BC100321.1 | - | 2 | 203 | |||
BC121824.1 | - | 2 | 220 | 40107149 |