Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177150.2 | NP_796124.1 | 11 | 1328 | ||
GenBank mRNA | AK089172.1 | - | 11 | 1377 | ||
AK090340.1 | - | 11 | 1329 | |||
AK143613.1 | - | 11 | 1291 | |||
AK164039.1 | - | 7 | 929 | |||
AK167376.1 | - | 11 | 1329 | |||
BC022690.1 | - | 2 | 188 | |||
BC080832.1 | - | 10 | 1319 | |||
BC100321.1 | - | 5 | 2759 | |||
BC121824.1 | - | 11 | 1329 | 40107149 |