Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177152.5 | NP_796126.4 | 6 | 1024 | ||
GenBank mRNA | AK035166.1 | - | 6 | 1026 | ||
AK137007.1 | - | 6 | 1011 | |||
AY505341.1 | - | 6 | 967 | |||
BC065142.1 | - | 6 | 990 | 30610561 |