Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klri2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177155.4 | NP_796129.2 | 6 | 1040 | ||
GenBank mRNA | AK040915.1 | - | 6 | 1027 | ||
AK041213.1 | - | 6 | 1043 | |||
BC132406.1 | - | 6 | 1020 | 40130683 | ||
BC132408.1 | - | 6 | 1020 | 40130685 |