Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9930014A18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035741.1 | - | 1 | 1269 | ||
AK036813.1 | - | 2 | 2668 | |||
BC099672.1 | - | 1 | 526 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177166.1 | - | 1269 | Chr. - - [-] on Build |