Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A830018L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160369.2 | NP_001153841.1 | 3 | 702 | ||
NM_001160370.2 | NP_001153842.1 | 3 | 702 | |||
NM_001160371.2 | NP_001153843.1 | 3 | 702 | |||
NM_177173.6 | NP_796147.2 | 3 | 702 | |||
GenBank mRNA | AK034391.1 | - | 3 | 698 | ||
AK039555.1 | - | 3 | 509 | |||
AK043673.1 | - | 3 | 539 | |||
AK081622.1 | - | 3 | 703 | |||
AK135047.1 | - | 3 | 696 | |||
AK136763.1 | - | 3 | 503 | |||
BC145022.1 | - | 3 | 298 | 9053570 |