Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166009.1 | NP_001159481.1 | 2 | 1158 | ||
NM_177175.4 | NP_796149.2 | 2 | 2100 | |||
GenBank mRNA | AK044217.1 | - | 2 | 2102 | ||
AK044627.1 | - | 2 | 2102 | |||
BC144984.1 | - | 2 | 1155 | 9053532 | ||
BC148227.1 | - | 2 | 1232 | 40140720 | ||
BC148284.1 | - | 2 | 1232 | 40140721 |