Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130105.1 | NP_001123577.1 | 13, 14 | 1802 | ||
NM_005713.2 | NP_005704.1 | 12, 13 | 1703 | |||
NM_031361.2 | NP_112729.1 | 11, 12 | 1625 | |||
XM_006714513.2 | XP_006714576.1 | 12, 13 | 1703 | |||
XM_011543090.2 | XP_011541392.1 | 12, 13 | 1703 | |||
XM_011543091.2 | XP_011541393.1 | 11, 12 | 1625 | |||
XM_017008919.1 | XP_016864408.1 | 11, 12 | 1625 | |||
GenBank mRNA | AF136450.1 | - | 12, 13 | 1689 | ||
AF232930.1 | - | 11, 12 | 1593 | |||
AK000604.1 | - | 7, 8 | 645 | |||
AK091851.1 | - | 12, 13 | 1515 | |||
AK096854.1 | - | 13, 14 | 1959 | |||
AK292087.1 | - | 11, 12 | 1625 | |||
AY453385.1 | - | 11, 12 | 1203 | |||
AY453386.1 | - | 12, 13 | 1281 | |||
BC000102.2 | - | 11, 12 | 1609 | 3505746 |