Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113354.1 | NP_001106825.1 | 4 | 1076 | ||
NM_177201.5 | NP_796175.1 | 4 | 1076 | |||
GenBank mRNA | AK036609.1 | - | 4 | 450 | ||
AK040943.1 | - | 4 | 445 | |||
AK040969.1 | - | 4 | 1077 | |||
AK122447.1 | - | 3 | 167 | |||
AK134137.1 | - | 4 | 413 | |||
AK147939.1 | - | 4 | 463 | |||
BC138899.1 | - | 4 | 345 | 8861921 |