Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177213.3 | NP_796187.2 | 3 | 283 | ||
GenBank mRNA | AK029712.1 | - | 3 | 283 | ||
AK076666.1 | - | 3 | 290 | |||
AY243471.1 | - | 3 | 283 | |||
BC089470.1 | - | 3 | 297 | |||
BC141350.1 | - | 3 | 248 | 9055962 | ||
BC141351.1 | - | 3 | 248 | 9055963 |