Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ocrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177215.3 | NP_796189.2 | 14 | 1629 | ||
GenBank mRNA | AK035315.1 | - | 14 | 1632 | ||
AK041963.1 | - | 14 | 1631 | |||
AK044320.1 | - | 13 | 1551 | |||
AK047906.1 | - | 14 | 1648 | |||
BC043709.1 | - | 11 | ||||
BC068146.1 | - | 14 | 1629 | 30533468 |