Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mysm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177239.2 | NP_796213.2 | 6 | 378 | ||
GenBank mRNA | AK043802.1 | - | 6 | 378 | ||
AK081684.1 | - | 6 | 353 | |||
AK164103.1 | - | 6 | 324 | |||
AK173300.1 | - | 3 | 120 | |||
BC150946.1 | - | 6 | 374 | 9087857 | ||
BC151172.1 | - | 6 | 374 | 9088061 |