Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005723.3 | NP_005714.2 | 7 | 1133 | ||
XM_005262680.1 | XP_005262737.1 | 6 | 986 | |||
GenBank mRNA | AF065389.1 | - | 7 | 1045 | ||
AK126702.1 | - | 4 | 3772 | |||
AK295385.1 | - | 7 | 727 | |||
AK315723.1 | - | 7 | 1100 | |||
BC009704.1 | - | 7 | 1031 | 3895933 | ||
CR541809.1 | - | 7 | 698 | |||
CR541829.1 | - | 7 | 698 |