Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168412.1 | NP_001161884.1 | 6 | 1468 | ||
NM_005724.5 | NP_005715.1 | 7 | 1660 | |||
NM_198902.2 | NP_944492.1 | 6 | 1585 | |||
XM_017021857.1 | XP_016877346.1 | 7 | 1269 | |||
GenBank mRNA | AF133423.1 | - | 7 | 1461 | ||
AK001305.1 | - | 7 | 1506 | |||
AK001310.1 | - | 7 | 1492 | |||
AK001326.1 | - | 7 | 1603 | |||
AK027793.1 | - | 4 | 2711 | |||
AK075336.1 | - | 7 | 1541 | |||
AK075482.1 | - | 6 | 1466 | |||
AK298157.1 | - | 6 | 1349 | |||
BC000704.2 | - | 7 | 1561 | 3349181 | ||
BC004280.1 | - | 7 | 1522 | 3616380 | ||
BC009248.2 | - | 7 | 1525 | 2964568 | ||
BC011206.1 | - | 7 | 1554 | 3457158 | ||
M69023.1 | - | 3 | 935 |
Gene Symbol: ANKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK222631.1 | - | 1775 |