Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005727.3 | NP_005718.2 | 9 | 1600 | ||
GenBank mRNA | AF065388.1 | - | 8 | 1247 | ||
BC007290.1 | - | 8 | 1253 | 3349295 | ||
BC013404.2 | - | 8 | 1251 | 2959379 | ||
BM856468.1 | - | 1 | 235 |