Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026509.1 | - | 2 | 158 | ||
BU660651.1 | - | 2 | 102 | |||
L20320.1 | - | 2 | 180 | |||
X77303.1 | - | 2 | 135 | |||
X77743.1 | - | 2 | 126 | |||
X79193.1 | - | 2 | 182 | |||
Y13120.1 | - | 2 | 195 |