Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005736.3 | NP_005727.1 | 11 | 2293 | ||
GenBank mRNA | AK096085.1 | - | 4 | 2939 | ||
BC000693.2 | - | 11 | 2229 | 3347881 | ||
BC026016.2 | - | 11 | 2195 | 3009822 | ||
X82206.1 | - | 11 | 2249 |