Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177305.4 | NP_796279.2 | 1 | 643 | ||
GenBank mRNA | AK080335.1 | - | 1 | 587 | ||
AK080380.1 | - | 1 | 587 | |||
AK080743.1 | - | 1 | 596 | |||
AK169890.1 | - | 1 | 80 | |||
BC049804.1 | - | 328 | ||||
BC055769.1 | - | 1 | 603 | 6414771 |